Product Details

SNP ID
rs4825424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120616496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAGTAAAGGACCACAACTCCAC[A/G]AGATCAGGAATGAATGAAGAAAATT
Phenotype
MIM: 300611 MIM: 300587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1GALT1C1 PubMed Links

Gene Details

Gene
C1GALT1C1
Gene Name
C1GALT1 specific chaperone 1
There are no transcripts associated with this gene.

Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137554.1 4938 UTR 3 NP_001131026.1
NM_014060.2 4938 UTR 3 NP_054779.1

View Full Product Details