Product Details

SNP ID
rs9463517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49607428 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTAGAAAATTCTGAGATACACCT[C/T]AAAGAAGAGAATCATCTTTTCATGT
Phenotype
MIM: 180297
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHAG PubMed Links

Gene Details

Gene
RHAG
Gene Name
Rh-associated glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000324.2 Intron NP_000315.2
XM_011514788.1 Intron XP_011513090.1

View Full Product Details