Product Details
- SNP ID
-
rs6038524
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:143570 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGATTATTTTTTCTAATGTGTGA[A/G]TACTGTAAATAACATTGTACAACCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB126
PubMed Links
Gene Details
- Gene
- DEFB126
- Gene Name
- defensin beta 126
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030931.3 |
|
Intron |
|
|
NP_112193.1 |
View Full Product Details