Product Details

SNP ID
rs6121907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62139295 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATACGAACATTTTAAACCATGC[C/T]TGAGCCCCTTCTCAGGTTATCAGCA
Phenotype
MIM: 606607 MIM: 606472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSM14B PubMed Links

Gene Details

Gene
LSM14B
Gene Name
LSM family member 14B
There are no transcripts associated with this gene.

Gene
PSMA7
Gene Name
proteasome subunit alpha 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002792.3 Intron NP_002783.1
Gene
SS18L1
Gene Name
SS18L1, nBAF chromatin remodeling complex subunit
There are no transcripts associated with this gene.

View Full Product Details