Product Details

SNP ID
rs6057583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32447366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCAGAGCTGCCCCGTTGGCCT[C/T]TTCCAAGCAGGTCAGAGCACCCGTG
Phenotype
MIM: 612990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASXL1 PubMed Links

Gene Details

Gene
ASXL1
Gene Name
additional sex combs like 1, transcriptional regulator
There are no transcripts associated with this gene.

Gene
NOL4L
Gene Name
nucleolar protein 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256798.1 1795 UTR 3 NP_001243727.1
NM_080616.4 1795 UTR 3 NP_542183.2
XM_005260282.4 1795 Intron XP_005260339.1
XM_005260283.4 1795 Intron XP_005260340.1
XM_005260284.4 1795 Intron XP_005260341.1
XM_005260285.4 1795 Intron XP_005260342.2
XM_005260286.4 1795 Intron XP_005260343.1
XM_005260288.2 1795 Intron XP_005260345.1
XM_005260289.4 1795 Intron XP_005260346.1
XM_006723702.3 1795 Intron XP_006723765.1
XM_006723703.3 1795 Intron XP_006723766.1
XM_011528562.2 1795 Intron XP_011526864.1
XM_011528563.1 1795 Intron XP_011526865.1
XM_017027669.1 1795 Intron XP_016883158.1
XM_017027670.1 1795 Intron XP_016883159.1

View Full Product Details