Product Details

SNP ID
rs10410188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56846653 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATTTCTACATAAGCATATATTT[C/T]TTTAACAGACTATAAAGTACTAAAG
Phenotype
MIM: 601483
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIMT1 PubMed Links

Gene Details

Gene
MIMT1
Gene Name
MER1 repeat containing imprinted transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
PEG3
Gene Name
paternally expressed 3
There are no transcripts associated with this gene.

Gene
ZIM2
Gene Name
zinc finger imprinted 2
There are no transcripts associated with this gene.

View Full Product Details