Product Details

SNP ID
rs9852462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32703623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATCTTATACAACATGGAATGCT[A/G]GACAAAGGAATGATTGACTTCCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT10 PubMed Links

Gene Details

Gene
CNOT10
Gene Name
CCR4-NOT transcription complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256741.1 Intron NP_001243670.1
NM_001256742.1 Intron NP_001243671.1
NM_015442.2 Intron NP_056257.1
XM_006713084.3 Intron XP_006713147.1
XM_006713085.3 Intron XP_006713148.1
XM_011533566.2 Intron XP_011531868.1
XM_011533567.2 Intron XP_011531869.1
XM_017006109.1 Intron XP_016861598.1
XM_017006110.1 Intron XP_016861599.1
XM_017006111.1 Intron XP_016861600.1
XM_017006112.1 Intron XP_016861601.1

View Full Product Details