Product Details

SNP ID
rs10032507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:173371807 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCGACCGGGACACGAAACAAAG[A/G]GGACCGGCGCACTGAGGGCCCAGAG
Phenotype
MIM: 603378
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAP30 PubMed Links

Gene Details

Gene
SAP30
Gene Name
Sin3A associated protein 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003864.3 Intron NP_003855.1

View Full Product Details