Product Details
- SNP ID
-
rs4830986
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:15688395 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCATGCATTTTACTGTTATCACAG[C/G]ACTGATTGCACTTGGTTTAGGTGAG
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CA5BP1
PubMed Links
Gene Details
- Gene
- CA5BP1
- Gene Name
- carbonic anhydrase 5B pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details