Product Details
- SNP ID
-
rs10424448
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2904860 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACCTGGCCTCCCTCGTAGCTTACT[C/G]TCTGTGTGTGGGCATATGTGAGCAC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF57
PubMed Links
Gene Details
- Gene
- ZNF57
- Gene Name
- zinc finger protein 57
View Full Product Details