Product Details
- SNP ID
-
rs5753184
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:30529852 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCTCCCACGTGAGTCAGACAATA[C/G]TTCAGCTTCAGGAATGAGAGAGACT
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SEC14L6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114264860] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEC14L6
- Gene Name
- SEC14 like lipid binding 6
There are no transcripts associated with this gene.
View Full Product Details