Product Details

SNP ID
rs10230212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75932002 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTGCTGCTAGGCCTCTGTGCACA[C/T]AGTGCTTTGTGGAATTGCTCCCGTC
Phenotype
MIM: 124015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POR PubMed Links

Gene Details

Gene
POR
Gene Name
cytochrome p450 oxidoreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000941.2 Intron NP_000932.3

View Full Product Details