Product Details

SNP ID
rs6119479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34255244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCTTGGATTTGTGAATATTGAA[C/G]TGGACTGTTGATTCAGCCCCCTCGT
Phenotype
MIM: 180960 MIM: 600201
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AHCY PubMed Links

Gene Details

Gene
AHCY
Gene Name
adenosylhomocysteinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000687.3 Intron NP_000678.1
NM_001161766.1 Intron NP_001155238.1
NM_001322084.1 Intron NP_001309013.1
NM_001322085.1 Intron NP_001309014.1
NM_001322086.1 Intron NP_001309015.1
XM_005260317.2 Intron XP_005260374.1
XM_011528656.2 Intron XP_011526958.2
XM_011528657.2 Intron XP_011526959.2
XM_011528658.2 Intron XP_011526960.2
XM_011528659.1 Intron XP_011526961.1
XM_017027708.1 Intron XP_016883197.1
XM_017027709.1 Intron XP_016883198.1
XM_017027710.1 Intron XP_016883199.1
Gene
ASIP
Gene Name
agouti signaling protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001672.2 Intron NP_001663.2
XM_005260412.3 Intron XP_005260469.1
XM_011528820.2 Intron XP_011527122.1
XM_011528821.1 Intron XP_011527123.1

View Full Product Details