Product Details
- SNP ID
-
rs6640588
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:10020599 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCCAGATCCTTTTGCTAATGGTGT[C/G]TATCTTTCCCATAAATTCAATAAAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WWC3
PubMed Links
Gene Details
- Gene
- WWC3
- Gene Name
- WWC family member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015691.3 |
|
Intron |
|
|
NP_056506.2 |
View Full Product Details