Product Details

SNP ID
rs6121012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35652561 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTTTTACTTTTCTTGAACCTATA[A/G]GCCTGTCATTTAAGTCAATGACAGC
Phenotype
MIM: 604205 MIM: 607179
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPNE1 PubMed Links

Gene Details

Gene
CPNE1
Gene Name
copine 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198863.1 3034 Intron NP_001185792.1
NM_003915.5 3034 Intron NP_003906.2
NM_152925.2 3034 Intron NP_690902.1
NM_152926.2 3034 Intron NP_690903.1
NM_152927.2 3034 Intron NP_690904.1
NM_152928.2 3034 Intron NP_690905.1
Gene
RBM12
Gene Name
RNA binding motif protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198838.1 3034 Missense Mutation CCT,CTT P921L NP_001185767.1
NM_001198840.1 3034 Missense Mutation CCT,CTT P921L NP_001185769.1
NM_006047.5 3034 Missense Mutation CCT,CTT P921L NP_006038.2
NM_152838.3 3034 Missense Mutation CCT,CTT P921L NP_690051.1

View Full Product Details