Product Details

SNP ID
rs6110018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1442922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCCTGATTCTGAACTCACTTA[A/G]GATGGGTTGGCATGAGCTGTATCCT
Phenotype
MIM: 606610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSFL1C PubMed Links

Gene Details

Gene
NSFL1C
Gene Name
NSFL1 cofactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206736.1 3365 UTR 3 NP_001193665.1
NM_016143.4 3365 UTR 3 NP_057227.2
NM_018839.4 3365 UTR 3 NP_061327.2
XM_006723592.2 3365 UTR 3 XP_006723655.1
XM_006723593.2 3365 UTR 3 XP_006723656.1
XM_006723594.2 3365 UTR 3 XP_006723657.1
XM_011529300.1 3365 UTR 3 XP_011527602.1
XM_011529301.1 3365 UTR 3 XP_011527603.1
XM_011529302.2 3365 UTR 3 XP_011527604.1
XM_011529304.1 3365 UTR 3 XP_011527606.1
XM_011529305.2 3365 Intron XP_011527607.1
XM_017027956.1 3365 UTR 3 XP_016883445.1
XM_017027957.1 3365 UTR 3 XP_016883446.1
XM_017027958.1 3365 UTR 3 XP_016883447.1

View Full Product Details