Product Details
- SNP ID
-
rs5959652
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:80018000 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATCCTCCCCATGAGAAGAACACAG[C/T]GAACACAGATAAGAATTTGTATCTC
- Phenotype
-
MIM: 300307
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TBX22
PubMed Links
Gene Details
- Gene
- TBX22
- Gene Name
- T-box 22
There are no transcripts associated with this gene.
View Full Product Details