Product Details

SNP ID
rs9948906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:21826269 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACTGCTTCTTTTAGCATTATAG[C/T]TGAGGGCTAATAGCAAATTGTTGAT
Phenotype
MIM: 608677 MIM: 610252 MIM: 610254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIB1 PubMed Links

Gene Details

Gene
MIB1
Gene Name
mindbomb E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020774.3 Intron NP_065825.1
XM_011526098.1 Intron XP_011524400.1
XM_017025873.1 Intron XP_016881362.1
XM_017025874.1 Intron XP_016881363.1
XM_017025875.1 Intron XP_016881364.1
Gene
MIR1-2
Gene Name
microRNA 1-2
There are no transcripts associated with this gene.

Gene
MIR133A1
Gene Name
microRNA 133a-1
There are no transcripts associated with this gene.

Gene
MIR133A1HG
Gene Name
MIR133A1 host gene
There are no transcripts associated with this gene.

View Full Product Details