Product Details

SNP ID
rs10257050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95583892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCTACTGCAAACTGTTTTGTAT[A/G]ATCAGAAATGCATGAGCTGGACTCC
Phenotype
MIM: 602527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDK4 PubMed Links

Gene Details

Gene
PDK4
Gene Name
pyruvate dehydrogenase kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002612.3 3305 UTR 3 NP_002603.1

View Full Product Details