Product Details

SNP ID
rs4952590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:42049781 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACGTCCTCTCTCTCAGGTCTCCC[C/T]CCATCCTAAGATTAGACAGACTATT
Phenotype
MIM: 614150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKDCC PubMed Links

Gene Details

Gene
PKDCC
Gene Name
protein kinase domain containing, cytoplasmic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138370.2 Intron NP_612379.2

View Full Product Details