Product Details
- SNP ID
-
rs7552416
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
24
- Location
-
Chr.1:152842930 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGGTAAGGCTACTTCTGGCTGAG[A/G]AGACACTTGGATGTAGCTCAAGTGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LCE6A
PubMed Links
Gene Details
- Gene
- LCE6A
- Gene Name
- late cornified envelope 6A
View Full Product Details