Product Details

SNP ID
rs10057302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157087601 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATCAAGATGTCTGCAATTATGTA[A/C]ACATTTAGATTTTCAAAAAATGGGC
Phenotype
MIM: 606652
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAVCR2 PubMed Links

Gene Details

Gene
HAVCR2
Gene Name
hepatitis A virus cellular receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032782.4 Intron NP_116171.3

View Full Product Details