Product Details

SNP ID
rs10517375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:36288800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTACTTCATCTGTCAGTTGAGAAAC[A/G]TAACTCCTGGAAGAAAAGTGTATTC
Phenotype
MIM: 616979
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTHD1 PubMed Links

Gene Details

Gene
DTHD1
Gene Name
death domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136536.4 Intron NP_001130008.2
NM_001170700.2 Intron NP_001164171.1
XM_006714014.3 Intron XP_006714077.1
XM_011513693.2 Intron XP_011511995.1
XM_011513694.2 Intron XP_011511996.1
XM_011513695.2 Intron XP_011511997.1
XM_011513696.2 Intron XP_011511998.1
XM_017008191.1 Intron XP_016863680.1
XM_017008192.1 Intron XP_016863681.1
XM_017008193.1 Intron XP_016863682.1

View Full Product Details