Product Details

SNP ID
rs6413466
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:159712635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCAGATAGGGAGCTGGGGAGAT[A/G]GGAGATGGGCTCCTCTGACAGGACA
Phenotype
MIM: 123260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRP PubMed Links

Gene Details

Gene
CRP
Gene Name
C-reactive protein, pentraxin-related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000567.2 1669 UTR 3 NP_000558.2
XM_011509207.2 1669 UTR 3 XP_011507509.1

View Full Product Details