Product Details

SNP ID
rs9469333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33067172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGGAATGATCCAGGTATCCCAG[C/G]AGCAATGTAGATATGTGGATTAATA
Phenotype
MIM: 142880 MIM: 142858
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HLA-DPA1 PubMed Links

Gene Details

Gene
HLA-DPA1
Gene Name
major histocompatibility complex, class II, DP alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242524.1 Intron NP_001229453.1
NM_001242525.1 Intron NP_001229454.1
NM_033554.3 Intron NP_291032.2
Gene
HLA-DPB1
Gene Name
major histocompatibility complex, class II, DP beta 1
There are no transcripts associated with this gene.

View Full Product Details