Product Details

SNP ID
rs9358943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26462649 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCAGGCTAGATCTGTCAAGTCT[C/T]CTAAAAACCATCACCTCCTATGGGT
Phenotype
MIM: 613590 MIM: 613595
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTN2A1 PubMed Links

Gene Details

Gene
BTN2A1
Gene Name
butyrophilin subfamily 2 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197233.2 Intron NP_001184162.1
NM_001197234.2 Intron NP_001184163.1
NM_007049.4 Intron NP_008980.1
NM_078476.3 Intron NP_510961.1
Gene
BTN3A3
Gene Name
butyrophilin subfamily 3 member A3
There are no transcripts associated with this gene.

Gene
LOC285819
Gene Name
uncharacterized LOC285819
There are no transcripts associated with this gene.

View Full Product Details