Product Details

SNP ID
rs10416062
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42438000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTTTTCTACACATATACACTTA[C/T]GATAAAGTCTAATTTATGAATTAGG
Phenotype
MIM: 611387
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCL17 PubMed Links

Gene Details

Gene
CXCL17
Gene Name
C-X-C motif chemokine ligand 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198477.2 Intron NP_940879.1
Gene
LIPE-AS1
Gene Name
LIPE antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details