Product Details

SNP ID
rs9321504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:135504563 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGCTTTTTATGTCAGTAACATA[C/T]ATCTACCCCATTCATTTTAATGGCT
Phenotype
MIM: 608894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHI1 PubMed Links

Gene Details

Gene
AHI1
Gene Name
Abelson helper integration site 1
There are no transcripts associated with this gene.

Gene
LINC00271
Gene Name
long intergenic non-protein coding RNA 271
There are no transcripts associated with this gene.

View Full Product Details