Product Details

SNP ID
rs9679171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86200365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATGCATGAATGATTTAACAGTT[C/T]TAAAGCAAACACCTGTTTAACACTC
Phenotype
MIM: 600378 MIM: 611841
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMMT PubMed Links

Gene Details

Gene
IMMT
Gene Name
inner membrane mitochondrial protein
There are no transcripts associated with this gene.

Gene
MIR4779
Gene Name
microRNA 4779
There are no transcripts associated with this gene.

Gene
MRPL35
Gene Name
mitochondrial ribosomal protein L35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016622.3 Intron NP_057706.2
NM_145644.2 Intron NP_663619.1
XM_005264361.2 Intron XP_005264418.1

View Full Product Details