Product Details
- SNP ID
-
rs10173155
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:86026458 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCTGGTTTTACAAAGCCTTTGTA[C/T]AAGTCCTCTCTGTCTTTTCTGTCCA
- Phenotype
-
MIM: 616404
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC90784
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77962929] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC90784
- Gene Name
- uncharacterized LOC90784
There are no transcripts associated with this gene.
- Gene
- POLR1A
- Gene Name
- RNA polymerase I subunit A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015425.4 |
6507 |
UTR 3 |
|
|
NP_056240.2 |
View Full Product Details