Product Details

SNP ID
rs10229931
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30777480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCTTTCCATTTCCCTTCCCTA[C/T]GGCCTCCTCTTTCCTCTGTTTGTTT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM188B PubMed Links

Gene Details

Gene
FAM188B
Gene Name
family with sequence similarity 188 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032222.2 Intron NP_115598.2
Gene
INMT-FAM188B
Gene Name
INMT-FAM188B readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details