Product Details

SNP ID
rs6638297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134892967 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTCTGGCTCTATGTGTATAGTA[C/T]ACATACTCATATTTTTAAGAAACCC
Phenotype
MIM: 300674
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOSPD1 PubMed Links

Gene Details

Gene
MOSPD1
Gene Name
motile sperm domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306188.1 Intron NP_001293117.1
NM_019556.2 Intron NP_062456.1
XM_011531366.2 Intron XP_011529668.1
XM_017029653.1 Intron XP_016885142.1
XM_017029654.1 Intron XP_016885143.1

View Full Product Details