Product Details

SNP ID
rs6037055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:25199007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTCCTGTGGCATCTATGATGC[C/T]TGCATCTTCCTCCTACTGCCCGTTT
Phenotype
MIM: 603160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENTPD6 PubMed Links

Gene Details

Gene
ENTPD6
Gene Name
ectonucleoside triphosphate diphosphohydrolase 6 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114089.3 Intron NP_001107561.1
NM_001247.4 Intron NP_001238.2
NM_001317941.2 Intron NP_001304870.1
NM_001322378.1 Intron NP_001309307.1
NM_001322379.1 Intron NP_001309308.1
NM_001322380.1 Intron NP_001309309.1
NM_001322381.1 Intron NP_001309310.1
NM_001322382.1 Intron NP_001309311.1
NM_001322383.1 Intron NP_001309312.1
NM_001322384.1 Intron NP_001309313.1
NM_001322385.1 Intron NP_001309314.1
NM_001322386.1 Intron NP_001309315.1
NM_001322387.1 Intron NP_001309316.1
NM_001322388.1 Intron NP_001309317.1
NM_001322389.1 Intron NP_001309318.1
NM_001322390.1 Intron NP_001309319.1
NM_001322391.1 Intron NP_001309320.1
NM_001322392.1 Intron NP_001309321.1
NM_001322393.1 Intron NP_001309322.1
NM_001322394.1 Intron NP_001309323.1
NM_001322395.1 Intron NP_001309324.1
NM_001322396.1 Intron NP_001309325.1
NM_001322397.1 Intron NP_001309326.1
NM_001322398.1 Intron NP_001309327.1
XM_006723665.2 Intron XP_006723728.1
XM_011529396.1 Intron XP_011527698.1
XM_011529397.1 Intron XP_011527699.1
XM_017028129.1 Intron XP_016883618.1
XM_017028130.1 Intron XP_016883619.1
XM_017028131.1 Intron XP_016883620.1
XM_017028132.1 Intron XP_016883621.1
XM_017028133.1 Intron XP_016883622.1
XM_017028134.1 Intron XP_016883623.1
Gene
LOC101926889
Gene Name
uncharacterized LOC101926889
There are no transcripts associated with this gene.

View Full Product Details