Product Details

SNP ID
rs6074572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13232949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCTCATGGGTCTCGATTCTCCA[A/G]GTGATGGTGGTGACAGACATTGATT
Phenotype
MIM: 615793 MIM: 608270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISM1 PubMed Links

Gene Details

Gene
ISM1
Gene Name
isthmin 1, angiogenesis inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080826.1 Intron NP_543016.1
XM_017027680.1 Intron XP_016883169.1
Gene
ISM1-AS1
Gene Name
ISM1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TASP1
Gene Name
taspase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323602.1 Intron NP_001310531.1
NM_001323603.1 Intron NP_001310532.1
NM_001323604.1 Intron NP_001310533.1
NM_017714.2 Intron NP_060184.2
XM_011529292.1 Intron XP_011527594.1
XM_011529294.1 Intron XP_011527596.1
XM_017027927.1 Intron XP_016883416.1
XM_017027928.1 Intron XP_016883417.1
XM_017027929.1 Intron XP_016883418.1
XM_017027930.1 Intron XP_016883419.1
XM_017027931.1 Intron XP_016883420.1

View Full Product Details