Product Details

SNP ID
rs4927036
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:53782845 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATAGTTATCTTGAAATATCATTC[A/G]TCTTATCCCTACCTCAAAATAAGTT
Phenotype
MIM: 610115
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDC1 PubMed Links

Gene Details

Gene
NDC1
Gene Name
NDC1 transmembrane nucleoporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168551.1 Intron NP_001162023.1
NM_018087.4 Intron NP_060557.3
XM_011541766.2 Intron XP_011540068.1

View Full Product Details