Product Details

SNP ID
rs8182512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3575104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAAAGATATGTCTAAGAAGTCA[A/G]GATGGGGCCTGGATTTTTTGCGAAA
Phenotype
MIM: 605535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMG20B PubMed Links

Gene Details

Gene
HMG20B
Gene Name
high mobility group 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006339.2 Intron NP_006330.2
XM_017026144.1 Intron XP_016881633.1

View Full Product Details