Product Details

SNP ID
rs6819963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143581124 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCCTCCAGCCCAGTGCAGCAGG[A/C]TTATAGAAAACTGCCCAGACTGTTC
Phenotype
MIM: 608946
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FREM3 PubMed Links

Gene Details

Gene
FREM3
Gene Name
FRAS1 related extracellular matrix 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168235.1 Intron NP_001161707.1

View Full Product Details