Product Details

SNP ID
rs10217131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21809334 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTCAGCTTGCCACACTCAGCCCA[C/T]GTTTTAAAAATTTATGGTCTAGGGG
Phenotype
MIM: 156540
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTAP PubMed Links

Gene Details

Gene
MTAP
Gene Name
methylthioadenosine phosphorylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002451.3 Intron NP_002442.2

View Full Product Details