Product Details

SNP ID
rs10421208
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10853947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGAGGGACTGCTGGCATGTGTCA[A/G]TAGGCTGAGGCAGGTGGATCACTTA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C19orf38 PubMed Links
Additional Information
For this assay, SNP(s) [rs545079431] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C19orf38
Gene Name
chromosome 19 open reading frame 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136482.1 Intron NP_001129954.1
XM_005259846.4 Intron XP_005259903.1
XM_005259847.4 Intron XP_005259904.1

View Full Product Details