Product Details
- SNP ID
-
rs9834784
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:169971564 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTTGGGAGAGATTATCCAAGTCC[A/G]TGTTTTCTGGTCTCTTCCTTTCATT
- Phenotype
-
MIM: 602173
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100128164
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114081959] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100128164
- Gene Name
- four and a half LIM domains 1 pseudogene
There are no transcripts associated with this gene.
- Gene
- SEC62
- Gene Name
- SEC62 homolog, preprotein translocation factor
View Full Product Details