Product Details

SNP ID
rs6642501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:111780453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTACCATTACCCTGTGTATTTTTT[C/G]TTCATGGCAGTTTTTATCCTTGTAA
Phenotype
MIM: 300334
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRPC5 PubMed Links

Gene Details

Gene
TRPC5
Gene Name
transient receptor potential cation channel subfamily C member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012471.2 Intron NP_036603.1
XM_017029774.1 Intron XP_016885263.1

View Full Product Details