Product Details

SNP ID
rs8177351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126283508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCACGTTCTTCTCTGCTCCGGT[C/G]ACAGCCTCTTCCTCTGCTCTTCGGG
Phenotype
MIM: 613622 MIM: 606252
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FOXRED1 PubMed Links

Gene Details

Gene
FOXRED1
Gene Name
FAD dependent oxidoreductase domain containing 1
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039661.1 Intron NP_001034750.1
NM_001318776.1 Intron NP_001305705.1
NM_001318777.1 Intron NP_001305706.1
NM_148910.2 Intron NP_683708.1
XM_005271399.3 Intron XP_005271456.2
XM_011542576.2 Intron XP_011540878.1
XM_017017161.1 Intron XP_016872650.1
XM_017017162.1 Intron XP_016872651.1

View Full Product Details