Product Details

SNP ID
rs6122201
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63870366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCTGAGGACCAGGTTTCAGATC[C/T]TGAGGACATCACACAGCTATTTCTA
Phenotype
MIM: 603747
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ABHD16B PubMed Links

Gene Details

Gene
ABHD16B
Gene Name
abhydrolase domain containing 16B
There are no transcripts associated with this gene.

Gene
TPD52L2
Gene Name
tumor protein D52 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243891.1 Intron NP_001230820.1
NM_001243892.1 Intron NP_001230821.1
NM_001243894.1 Intron NP_001230823.1
NM_001243895.1 Intron NP_001230824.1
NM_003288.3 Intron NP_003279.2
NM_199359.2 Intron NP_955391.1
NM_199360.2 Intron NP_955392.1
NM_199361.2 Intron NP_955393.1
NM_199362.2 Intron NP_955394.1
NM_199363.2 Intron NP_955395.1
XM_011529034.2 Intron XP_011527336.1
XM_011529035.2 Intron XP_011527337.1
XM_017028038.1 Intron XP_016883527.1

View Full Product Details