Product Details

SNP ID
rs5943453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:28593049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTACAAATATGCTTTTAATTTTCA[A/C]CCAGAAAATGCCACCCTGGAGTGTC
Phenotype
MIM: 300206
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IL1RAPL1 PubMed Links

Gene Details

Gene
IL1RAPL1
Gene Name
interleukin 1 receptor accessory protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014271.3 Intron NP_055086.1
XM_017029240.1 Intron XP_016884729.1
XM_017029241.1 Intron XP_016884730.1

View Full Product Details