Product Details
- SNP ID
-
rs10423962
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:55491914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCCGGCAGGCAGAGGGCACCCAC[C/T]TGGCATTTCATCAGCGTTGGGGTGG
- Phenotype
-
MIM: 610671
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NAT14
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77574931] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NAT14
- Gene Name
- N-acetyltransferase 14 (putative)
There are no transcripts associated with this gene.
- Gene
- SSC5D
- Gene Name
- scavenger receptor cysteine rich family member with 5 domains
- Gene
- ZNF628
- Gene Name
- zinc finger protein 628
There are no transcripts associated with this gene.
View Full Product Details