Product Details

SNP ID
rs7524082
Assay Type
Functionally tested
NCBI dbSNP Submissions
59
Location
Chr.1:27937570 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAGGACTGAAATAGAGACTGAGA[C/T]CATTGTGTGGTGTGGGTTATAGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMPDL3B PubMed Links

Gene Details

Gene
SMPDL3B
Gene Name
sphingomyelin phosphodiesterase acid like 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009568.2 Intron NP_001009568.1
NM_001304579.1 Intron NP_001291508.1
NM_014474.3 Intron NP_055289.2
XM_011541259.2 Intron XP_011539561.1
XM_011541260.2 Intron XP_011539562.1

View Full Product Details