Product Details

SNP ID
rs9904758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55269396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGATCCCCTGCTACTTAGAAT[A/G]TGCGGTACCACTATCTTCAATTTTT
Phenotype
MIM: 142385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLF PubMed Links

Gene Details

Gene
HLF
Gene Name
HLF, PAR bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002126.4 Intron NP_002117.1
XM_005257269.2 Intron XP_005257326.1
XM_011524705.1 Intron XP_011523007.1
XM_017024556.1 Intron XP_016880045.1

View Full Product Details