Product Details

SNP ID
rs4646318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19967324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAAGGCAGCCGCCCTGCTCAA[A/G]GCCTAGGCCATTGTCCTCCTCCCGG
Phenotype
MIM: 602269 MIM: 116790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARVCF PubMed Links

Gene Details

Gene
ARVCF
Gene Name
armadillo repeat gene deleted in velocardiofacial syndrome
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001670.2 Intron NP_001661.1
XM_005261242.2 Intron XP_005261299.1
XM_005261243.3 Intron XP_005261300.1
XM_005261244.3 Intron XP_005261301.1
XM_006724243.2 Intron XP_006724306.1
XM_006724245.2 Intron XP_006724308.1
XM_006724246.3 Intron XP_006724309.1
XM_006724247.3 Intron XP_006724310.1
XM_006724248.3 Intron XP_006724311.1
XM_006724249.3 Intron XP_006724312.1
XM_006724250.3 Intron XP_006724313.1
XM_011530179.2 Intron XP_011528481.1
XM_011530180.1 Intron XP_011528482.1
XM_011530181.1 Intron XP_011528483.1
XM_011530182.2 Intron XP_011528484.1
XM_011530183.2 Intron XP_011528485.1
Gene
COMT
Gene Name
catechol-O-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000754.3 Intron NP_000745.1
NM_001135161.1 Intron NP_001128633.1
NM_001135162.1 Intron NP_001128634.1
NM_007310.2 Intron NP_009294.1
XM_011529886.1 Intron XP_011528188.1
XM_017028594.1 Intron XP_016884083.1
XM_017028595.1 Intron XP_016884084.1
Gene
MIR4761
Gene Name
microRNA 4761
There are no transcripts associated with this gene.

View Full Product Details