Product Details
- SNP ID
-
rs5962321
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:106203435 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCTTATGGAATAAGGATGGAAA[C/G]TACATCTGATAGTGAGTGAAGTCCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MUM1L1
PubMed Links
Gene Details
- Gene
- MUM1L1
- Gene Name
- MUM1 like 1
View Full Product Details