Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286259.1 | 5836 | UTR 3 | NP_001273188.1 | ||
NM_182503.2 | 5836 | UTR 3 | NP_872309.2 | ||
XM_011535439.2 | 5836 | UTR 3 | XP_011533741.1 | ||
XM_011535440.1 | 5836 | UTR 3 | XP_011533742.1 | ||
XM_011535443.1 | 5836 | Intron | XP_011533745.1 | ||
XM_011535444.2 | 5836 | UTR 3 | XP_011533746.1 | ||
XM_017010260.1 | 5836 | UTR 3 | XP_016865749.1 | ||
XM_017010261.1 | 5836 | UTR 3 | XP_016865750.1 | ||
XM_017010262.1 | 5836 | UTR 3 | XP_016865751.1 | ||
XM_017010263.1 | 5836 | UTR 3 | XP_016865752.1 |